Assay Details
Gene Symbol: TCP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137683.2 | XP_014993169.1 | 767 | 134 | ||
XM_015137684.2 | XP_014993170.1 | 762 | 134 | |||
XM_015137685.2 | XP_014993171.1 | 781 | 134 | |||
XM_015137688.2 | XP_014993174.1 | 766 | 134 | |||
XM_015137689.2 | XP_014993175.2 | 767 | 134 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
