Assay Details
Gene Symbol: FAM18B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113308.4 | XP_001113308.3 | 103 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001113308.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
