Assay Details
Gene Symbol: LOC719436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124675.2 | XP_014980161.2 | 82 | 87 | ||
XM_015124676.2 | XP_014980162.2 | 82 | 87 | |||
XM_015124677.2 | XP_014980163.2 | 82 | 87 | |||
XM_028839831.1 | XP_028695664.1 | 47 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015124676.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
