Assay Details
Gene Symbol: SYCE1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126665.2 | XP_014982151.2 | 470 | 54 | ||
XM_028841438.1 | XP_028697271.1 | 1887 | 54 | |||
XM_028841439.1 | XP_028697272.1 | 365 | 54 | |||
XM_028841440.1 | XP_028697273.1 | 470 | 54 | |||
XM_028841441.1 | XP_028697274.1 | 1812 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
