Assay Details
Gene Symbol: SMIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125557.2 | XP_014981043.1 | 697 | 79 | ||
XM_015125558.2 | XP_014981044.1 | 692 | 79 | |||
XM_015125561.2 | XP_014981047.1 | 689 | 79 | |||
XM_028840628.1 | XP_028696461.1 | 359 | 79 | |||
XM_028840630.1 | XP_028696463.1 | 359 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
