Assay Details
| Gene Symbol: LOC711440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001104543.4 | XP_001104543.1 | 92 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001104543.4 transcripts.
Set Membership: |
|

LOC711440
Genomic Map