Assay Details
| Gene Symbol: DUSP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147243.2 | XP_015002729.2 | 100 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015147243.2 transcripts.
Set Membership: |
|

DUSP29
Genomic Map