Assay Details
| Gene Symbol: LOC711592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028828373.1 | XP_028684206.1 | 1204 | 111 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC711592| Gene Symbol: LOC711592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028828373.1 | XP_028684206.1 | 1204 | 111 | ||
Set Membership: |
|