Assay Details
| Gene Symbol: LOC707623 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849926.1 | XP_028705759.1 | 1259 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC707623| Gene Symbol: LOC707623 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849926.1 | XP_028705759.1 | 1259 | 100 | ||