Assay Details
Gene Symbol: LOC100428886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125184.2 | XP_014980670.2 | 334 | 66 |
Gene Symbol: HBZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028840872.1 | XP_028696705.1 | 520 | 66 | ||
XM_028840873.1 | XP_028696706.1 | 528 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
