Assay Details
Gene Symbol: WBP2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150483.2 | XP_015005969.2 | 1260 | 79 | ||
XM_015150484.2 | XP_015005970.2 | 1491 | 79 | |||
XM_015150485.2 | XP_015005971.2 | 1429 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
