Assay Details
| Gene Symbol: C2CD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110657.2 | XP_014966143.1 | 3808 | 82 | ||
| XM_015110658.2 | XP_014966144.1 | 4076 | 82 | |||
| XM_015110659.2 | XP_014966145.1 | 3870 | 82 | |||
More Information
Set Membership: |
|

C2CD6
Genomic Map