Assay Details
| Gene Symbol: C7H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144588.2 | XP_015000074.2 | 574 | 87 | ||
| XM_015144590.2 | XP_015000076.2 | 741 | 87 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7H14orf180| Gene Symbol: C7H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144588.2 | XP_015000074.2 | 574 | 87 | ||
| XM_015144590.2 | XP_015000076.2 | 741 | 87 | |||