Assay Details
| Gene Symbol: BRICD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084172.4 | XP_001084172.2 | 180 | 81 | ||
| XM_001084285.4 | XP_001084285.2 | 180 | 81 | |||
| XM_028841569.1 | XP_028697402.1 | 180 | 81 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001084172.4, XM_001084285.4, XM_028841569.1 transcripts.
Set Membership: |
|

BRICD5
Genomic Map