Assay Details
| Gene Symbol: ZNF91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123464.2 | XP_014978950.2 | 3474 | 92 | ||
| XM_028840094.1 | XP_028695927.1 | 3474 | 92 | |||
| XM_028840095.1 | XP_028695928.1 | 3474 | 92 | |||
| XM_028840096.1 | XP_028695929.1 | 3474 | 92 | |||
| XM_028840097.1 | XP_028695930.1 | 3474 | 92 | |||
| XM_028840099.1 | XP_028695932.1 | 3442 | 92 | |||
| XM_028840100.1 | XP_028695933.1 | 3786 | 92 | |||
More Information
Set Membership: |
|

ZNF91
Genomic Map