Assay Details
| Gene Symbol: NAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257641.1 | NP_001244570.1 | 1276 | 69 | ||
| XM_015110564.2 | XP_014966050.1 | 1758 | 69 | |||
| XM_015110566.2 | XP_014966052.1 | 1755 | 69 | |||
| XM_015110567.2 | XP_014966053.1 | 1712 | 69 | |||
| XM_015110568.2 | XP_014966054.1 | 1841 | 69 | |||
| XM_015110571.2 | XP_014966057.1 | 1559 | 69 | |||
| XM_028830354.1 | XP_028686187.1 | 1506 | 69 | |||
| XM_028830355.1 | XP_028686188.1 | 2011 | 69 | |||
| XM_028830356.1 | XP_028686189.1 | 2919 | 69 | |||
| XM_028830357.1 | XP_028686190.1 | 1980 | 69 | |||
| XM_028830358.1 | XP_028686191.1 | 1810 | 69 | |||
| XM_028830359.1 | XP_028686192.1 | 1353 | 69 | |||
| GenBank mRNA | JU334624.1 | - | 1276 | 69 | ||
| JV047137.1 | - | 1276 | 69 | |||
More Information
Set Membership: |
|

NAB1
Genomic Map