Assay Details
| Gene Symbol: S1PR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266185.1 | NP_001253114.1 | 917 | 114 | ||
| GenBank mRNA | JU473531.1 | - | 917 | 114 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

S1PR4| Gene Symbol: S1PR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266185.1 | NP_001253114.1 | 917 | 114 | ||
| GenBank mRNA | JU473531.1 | - | 917 | 114 | ||
Set Membership: |
|