Assay Details
| Gene Symbol: Vamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012663.2 | NP_036795.1 | 3 - 4 | 359 | 67 | |
| XM_006246593.2 | XP_006246655.1 | 3 - 4 | 777 | 67 | ||
| GenBank mRNA | AJ133104.1 | - | 3 - 4 | 343 | 67 | |
| BC074003.1 | - | 3 - 4 | 359 | 7097683 | 67 | |
| M24105.1 | - | 2 - 3 | 297 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vamp2
Genomic Map