Assay Details
| Gene Symbol: Pvrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012064.1 | NP_001012064.1 | 1 - 2 | 303 | 55 | |
| XM_006228421.1 | XP_006228483.1 | 1 - 2 | 459 | 55 | ||
| GenBank mRNA | BC090075.1 | - | 1 - 2 | 303 | 7189358 | 55 |
More Information
Updated Mapping Information
Set Membership: |
|

Pvrl2
Genomic Map