Assay Details
| Gene Symbol: RGD1305455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024969.2 | NP_001020140.2 | 2 - 3 | 663 | 72 | |
| GenBank mRNA | FQ211718.1 | - | 2 - 3 | 709 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1305455
Genomic Map