Assay Details
| Gene Symbol: Carm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030041.3 | NP_001025212.1 | 8 - 9 | 1117 | 77 | |
| NM_001034088.2 | NP_001029260.1 | 8 - 9 | 1117 | 77 | ||
| GenBank mRNA | AB201114.1 | - | 8 - 9 | 1117 | 77 | |
| AB201115.1 | - | 8 - 9 | 1117 | 77 | ||
| AB201116.1 | - | 8 - 9 | 1117 | 77 | ||
| AB201117.1 | - | 8 - 9 | 1117 | 77 | ||
| KJ672502.1 | - | 8 - 9 | 1027 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of growth plate cartilage chondrocyte proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
protein methylation
aging
positive regulation of cell proliferation
RNA splicing
pathogenesis
histone methylation
peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
intracellular steroid hormone receptor signaling pathway
intracellular estrogen receptor signaling pathway
negative regulation of protein binding
regulation of intracellular estrogen receptor signaling pathway
histone arginine methylation
histone H3-R2 methylation
histone H3-R17 methylation
positive regulation of fat cell differentiation
response to cAMP
endochondral bone morphogenesis
regulation of mRNA binding
negative regulation of dendrite development
transcription coactivator activity
protein binding
protein methyltransferase activity
histone-arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
ligand-dependent nuclear receptor transcription coactivator activity
protein-arginine omega-N asymmetric methyltransferase activity
histone methyltransferase activity (H3-R17 specific)
histone methyltransferase activity
protein homodimerization activity
transcription regulatory region DNA binding
lysine-acetylated histone binding
regulation of growth plate cartilage chondrocyte proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
protein methylation
aging
positive regulation of cell proliferation
RNA splicing
pathogenesis
histone methylation
peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
intracellular steroid hormone receptor signaling pathway
intracellular estrogen receptor signaling pathway
negative regulation of protein binding
regulation of intracellular estrogen receptor signaling pathway
histone arginine methylation
histone H3-R2 methylation
histone H3-R17 methylation
positive regulation of fat cell differentiation
response to cAMP
endochondral bone morphogenesis
regulation of mRNA binding
negative regulation of dendrite development
transcription coactivator activity
protein binding
protein methyltransferase activity
histone-arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
ligand-dependent nuclear receptor transcription coactivator activity
protein-arginine omega-N asymmetric methyltransferase activity
histone methyltransferase activity (H3-R17 specific)
histone methyltransferase activity
protein homodimerization activity
transcription regulatory region DNA binding
lysine-acetylated histone binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P325337 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P430345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53704 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P325337 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P430345 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P325337 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P430345 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P325337 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P430345 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P430345 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P325337 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24055 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18970 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 94444 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15366 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98564 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10908476 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908462 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908472 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908469 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10915568 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908478 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908465 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908467 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908466 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908474 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908477 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908464 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908475 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908463 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908470 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908461 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908471 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908468 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908473 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10915567 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10908460 | - | - |
| Rat Genome 230 2.0 Array | 1399048_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1399048_at | - | - |
| Affymetrix HT Rat Focus Array | 1399048_at | - | - |
| Affymetrix HT Rat Focus Array | 1399048_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800029711.rn.1 | TC0800003642.rn.1 | RTA TC |
| Rat Expression Set 230 | 1399048_at | - | - |
| Rat Expression Set 230 | 1399048_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908460 | - | - |
| Rat Genome U34 Set | rc_AI237700_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21871085 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375134 | - | - |

Carm1
Genomic Map