Assay Details
Gene Symbol: Kifc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198752.3 | NP_942047.2 | 12 - 13 | 1422 | 81 | |
GenBank mRNA | AJ250329.1 | - | 12 - 13 | 1422 | 81 | |
AJ250330.1 | - | 11 - 12 | 1463 | 81 | ||
FQ211833.1 | - | 10 - 11 | 1386 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
