Assay Details
Gene Symbol: Fosb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256509.1 | NP_001243438.1 | 1 - 2 | 451 | 85 | |
XM_008758883.1 | XP_008757105.1 | 1 - 2 | 965 | 85 | ||
GenBank mRNA | KC847115.1 | - | 1 - 2 | 744 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
