Assay Details
| Gene Symbol: Idh3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053581.1 | NP_446033.1 | 4 - 5 | 347 | 82 | |
| XM_006235048.2 | XP_006235110.1 | 4 - 5 | 384 | 82 | ||
| GenBank mRNA | BC079113.1 | - | 4 - 5 | 347 | 7126029 | 82 |
| FQ214820.1 | - | 4 - 5 | 345 | 82 | ||
| FQ215184.1 | - | 4 - 5 | 339 | 82 | ||
| FQ215350.1 | - | 4 - 5 | 347 | 82 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Idh3B
Genomic Map