Assay Details
| Gene Symbol: Oraov1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230782.1 | XP_006230844.1 | 2 - 3 | 237 | 73 | |
| XM_006230783.1 | XP_006230845.1 | 2 - 3 | 237 | 73 | ||
More Information
Set Membership: |
|

Oraov1
Genomic Map