Assay Details
| Gene Symbol: Gnb2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130734.2 | NP_570090.2 | 5 - 6 | 774 | 65 | |
| GenBank mRNA | BC063809.1 | - | 5 - 6 | 749 | 6919982 | 65 |
| FQ212638.1 | - | 5 - 6 | 745 | 65 | ||
| FQ213067.1 | - | 5 - 6 | 745 | 65 | ||
| FQ216167.1 | - | 5 - 6 | 745 | 65 | ||
| FQ218475.1 | - | 5 - 6 | 775 | 65 | ||
| FQ219492.1 | - | 5 - 6 | 746 | 65 | ||
| FQ219729.1 | - | 5 - 6 | 745 | 65 | ||
| FQ219900.1 | - | 5 - 6 | 745 | 65 | ||
| FQ220124.1 | - | 5 - 6 | 745 | 65 | ||
| FQ220151.1 | - | 5 - 6 | 745 | 65 | ||
| FQ220277.1 | - | 5 - 6 | 745 | 65 | ||
| FQ220314.1 | - | 5 - 6 | 745 | 65 | ||
| FQ220391.1 | - | 5 - 6 | 745 | 65 | ||
| FQ220419.1 | - | 5 - 6 | 745 | 65 | ||
| FQ220428.1 | - | 5 - 6 | 749 | 65 | ||
| FQ220430.1 | - | 5 - 6 | 744 | 65 | ||
| FQ220472.1 | - | 5 - 6 | 745 | 65 | ||
| FQ220494.1 | - | 5 - 6 | 745 | 65 | ||
| FQ220819.1 | - | 5 - 6 | 746 | 65 | ||
| FQ221075.1 | - | 5 - 6 | 65 | |||
| FQ221286.1 | - | 5 - 6 | 749 | 65 | ||
| FQ223855.1 | - | 5 - 6 | 744 | 65 | ||
| FQ224952.1 | - | 5 - 6 | 745 | 65 | ||
| FQ225047.1 | - | 5 - 6 | 749 | 65 | ||
| FQ225511.1 | - | 5 - 6 | 745 | 65 | ||
| FQ225843.1 | - | 5 - 6 | 775 | 65 | ||
| FQ226802.1 | - | 5 - 6 | 745 | 65 | ||
| FQ228007.1 | - | 5 - 6 | 749 | 65 | ||
| FQ229157.1 | - | 5 - 6 | 744 | 65 | ||
| FQ229247.1 | - | 5 - 6 | 745 | 65 | ||
| FQ229458.1 | - | 5 - 6 | 742 | 65 | ||
| FQ229569.1 | - | 5 - 6 | 746 | 65 | ||
| FQ229616.1 | - | 5 - 6 | 745 | 65 | ||
| FQ229633.1 | - | 5 - 6 | 745 | 65 | ||
| FQ229758.1 | - | 5 - 6 | 745 | 65 | ||
| FQ229908.1 | - | 5 - 6 | 746 | 65 | ||
| FQ229916.1 | - | 5 - 6 | 743 | 65 | ||
| FQ229923.1 | - | 5 - 6 | 744 | 65 | ||
| FQ230003.1 | - | 5 - 6 | 743 | 65 | ||
| FQ230117.1 | - | 5 - 6 | 745 | 65 | ||
| FQ230328.1 | - | 5 - 6 | 747 | 65 | ||
| FQ234000.1 | - | 5 - 6 | 751 | 65 | ||
| FQ234673.1 | - | 5 - 6 | 747 | 65 | ||
| FQ235255.1 | - | 5 - 6 | 745 | 65 | ||
| U03390.1 | - | 5 - 6 | 737 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gnb2l1
Genomic Map