Assay Details
| Gene Symbol: Car11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175708.3 | NP_783639.1 | 7 - 8 | 1024 | 68 | |
| GenBank mRNA | AY075025.1 | - | 7 - 8 | 808 | 68 | |
| BC086972.1 | - | 7 - 8 | 1024 | 7190978 | 68 | |
More Information
Set Membership: |
|

Car11
Genomic Map