Assay Details
Gene Symbol: Atf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108115.1 | NP_001101585.1 | 6 - 7 | 678 | 56 | |
XM_006242396.2 | XP_006242458.1 | 6 - 7 | 711 | 56 | ||
XM_008765746.1 | XP_008763968.1 | 5 - 6 | 2369 | 56 | ||
XM_008765747.1 | XP_008763969.1 | 6 - 7 | 896 | 56 | ||
XM_008765748.1 | XP_008763970.1 | 3 - 4 | 2511 | 56 | ||
GenBank mRNA | BC158605.1 | - | 757 | 7135404 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
