Assay Details
Gene Symbol: Cpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012533.1 | NP_036665.1 | 9 - 10 | 981 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P11449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113552 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11449 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11449 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P11449 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11449 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29552 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68419 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30463 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10822510 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822511 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822507 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822513 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822509 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822512 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822508 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822514 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822506 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822515 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10822505 | - | - |
Rat Genome 230 2.0 Array | 1370084_at | - | - |
Rat Genome 230 2.0 Array | 1370084_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370084_at | - | - |
Affymetrix HT Rat Focus Array | 1370084_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200022913.rn.1 | TC0200003353.rn.1 | RTA TC |
Rat Expression Set 230 | 1370084_at | - | - |
Rat Expression Set 230 | 1370084_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822505 | - | - |
Rat Genome U34 Set | rc_AI237825_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20789369 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351743 | - | - |
RatRef-12 Expression BeadChip | ILMN_65650 | - | - |
