Assay Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012557.2 | NP_036689.2 | 10 - 11 | 1251 | 71 | |
| XM_006253488.2 | XP_006253550.1 | 11 - 12 | 1526 | 71 | ||
| XM_006253489.2 | XP_006253551.1 | 10 - 11 | 1383 | 71 | ||
| XM_006253490.2 | XP_006253552.1 | 10 - 11 | 1228 | 71 | ||
| XM_006253491.2 | XP_006253553.1 | 7 - 8 | 2057 | 71 | ||
| XM_008771423.1 | XP_008769645.1 | 9 - 10 | 2373 | 71 | ||
| GenBank mRNA | BC168147.1 | - | 10 - 11 | 1251 | 9024689 | 71 |
| U73586.1 | - | 9 - 10 | 1027 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P247997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1418 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P247997 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P247997 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P247997 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P247997 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86238 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38771 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75900 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79819 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86230 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39633 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35068 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55756 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10793565 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793568 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793567 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793555 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793559 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793564 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793557 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793561 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793556 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793566 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793562 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793560 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793558 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793554 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793563 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10793553 | - | - |
| Rat Genome 230 2.0 Array | 1368616_at | - | - |
| Rat Genome 230 2.0 Array | 1368616_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368616_at | - | - |
| Affymetrix HT Rat Focus Array | 1368616_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700008042.rn.1 | TC1700001169.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368616_PM_at | - | - |
| Rat Expression Set 230 | 1368616_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793553 | - | - |
| Rat Genome U34 Set | U73586_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21494109 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358953 | - | - |
| RatRef-12 Expression BeadChip | ILMN_54510 | - | - |

Fancc
Genomic Map