Assay Details
Gene Symbol: Gad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012563.1 | NP_036695.1 | 4 - 5 | 595 | 79 | |
GenBank mRNA | M72422.1 | - | 4 - 5 | 595 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P554928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172470 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P554928 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P554928 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P554928 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P554928 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30736 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65726 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77983 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95700 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74734 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68653 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98413 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10796763 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796755 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796764 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796761 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796757 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796765 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796756 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796754 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796753 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796759 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796767 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796766 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796752 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796758 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796760 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796762 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10796751 | - | - |
Rat Genome 230 2.0 Array | 1369462_at | - | - |
Rat Genome 230 2.0 Array | 1369462_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369462_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369462_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700007855.rn.1 | TC1700001136.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700007851.rn.1 | TC1700001136.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700007857.rn.1 | TC1700001136.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700007853.rn.1 | TC1700001136.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700007850.rn.1 | TC1700001136.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700007854.rn.1 | TC1700001136.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700007858.rn.1 | TC1700001136.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700007852.rn.1 | TC1700001136.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700007856.rn.1 | TC1700001136.rn.1 | RTA TC |
Rat Expression Set 230 | 1369462_PM_at | - | - |
Rat Expression Set 230 | 1369462_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796751 | - | - |
Rat Genome U34 Set | M72422_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21268222 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371012 | - | - |
RatRef-12 Expression BeadChip | ILMN_66740 | - | - |
