Assay Details
| Gene Symbol: Glud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012570.2 | NP_036702.1 | 4 - 5 | 817 | 55 | |
| GenBank mRNA | BC081841.1 | - | 4 - 5 | 701 | 7107138 | 55 |
| X14044.1 | - | 4 - 5 | 707 | 55 | ||
| X14223.1 | - | 4 - 5 | 680 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glutamate biosynthetic process
glutamate catabolic process
glutamine metabolic process
long-term memory
response to aluminum ion
substantia nigra development
positive regulation of insulin secretion
oxidation-reduction process
tricarboxylic acid metabolic process
glutamate dehydrogenase (NAD+) activity
glutamate dehydrogenase [NAD(P)+] activity
ATP binding
GTP binding
enzyme binding
ADP binding
NAD+ binding
leucine binding
glutamate biosynthetic process
glutamate catabolic process
glutamine metabolic process
long-term memory
response to aluminum ion
substantia nigra development
positive regulation of insulin secretion
oxidation-reduction process
tricarboxylic acid metabolic process
glutamate dehydrogenase (NAD+) activity
glutamate dehydrogenase [NAD(P)+] activity
ATP binding
GTP binding
enzyme binding
ADP binding
NAD+ binding
leucine binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6239 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14157 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14157 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14157 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P259908 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14157 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P472500 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700483168 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702409504 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18563 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73878 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700483168 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 702409504 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10786977 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786978 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786979 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786972 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786973 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786975 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786982 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786976 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786971 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786974 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786981 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786970 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786980 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10786969 | - | - |
| Rat Genome 230 2.0 Array | 1387878_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370200_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387878_at | - | - |
| Rat Genome 230 2.0 Array | 1370200_at | - | - |
| Affymetrix HT Rat Focus Array | 1370200_at | - | - |
| Affymetrix HT Rat Focus Array | 1387878_at | - | - |
| Affymetrix HT Rat Focus Array | 1370200_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387878_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600001667.rn.1 | TC1600000147.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387878_PM_at | - | - |
| Rat Expression Set 230 | 1370200_at | - | - |
| Rat Expression Set 230 | 1370200_PM_at | - | - |
| Rat Expression Set 230 | 1387878_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786969 | - | - |
| Rat Genome U34 Set | rc_AI179613_at | - | - |
| Rat Genome U34 Set | rc_AI233216_at | - | - |
| Rat Genome U34 Set | rc_AI137421_at | - | - |
| Rat Genome U34 Set | rc_AI137421_at | - | - |
| Rat Genome U34 Set | rc_AI179613_at | - | - |
| Rat Genome U34 Set | rc_AI233216_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21586769 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358897 | - | - |
| RatRef-12 Expression BeadChip | ILMN_59253 | - | - |

Glud1
Genomic Map