Assay Details
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270854.1 | NP_001257783.1 | 7 - 8 | 785 | 78 | |
NM_012672.3 | NP_036804.2 | 8 - 9 | 893 | 78 | ||
XM_006251709.2 | XP_006251771.1 | 3 - 4 | 806 | 78 | ||
XM_008770500.1 | XP_008768722.1 | 5 - 6 | 697 | 78 | ||
GenBank mRNA | DQ191165.1 | - | 5 - 6 | 640 | 78 | |
HM043807.1 | - | 4 - 5 | 682 | 78 | ||
J03819.1 | - | 7 - 8 | 781 | 78 | ||
J03933.1 | - | 7 - 8 | 782 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
sensory perception of sound
negative regulation of female receptivity
regulation of heart contraction
female courtship behavior
positive regulation of cell proliferation
organ morphogenesis
regulation of lipid metabolic process
intracellular receptor signaling pathway
positive regulation of chondrocyte differentiation
steroid hormone mediated signaling pathway
positive regulation of ossification
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic organ development
Type I pneumocyte differentiation
regulation of cholesterol metabolic process
regulation of triglyceride metabolic process
double-stranded DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
thyroid hormone receptor activity
zinc ion binding
enzyme binding
chromatin DNA binding
protein homodimerization activity
sequence-specific DNA binding
thyroid hormone binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
sensory perception of sound
negative regulation of female receptivity
regulation of heart contraction
female courtship behavior
positive regulation of cell proliferation
organ morphogenesis
regulation of lipid metabolic process
intracellular receptor signaling pathway
positive regulation of chondrocyte differentiation
steroid hormone mediated signaling pathway
positive regulation of ossification
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic organ development
Type I pneumocyte differentiation
regulation of cholesterol metabolic process
regulation of triglyceride metabolic process
double-stranded DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
thyroid hormone receptor activity
zinc ion binding
enzyme binding
chromatin DNA binding
protein homodimerization activity
sequence-specific DNA binding
thyroid hormone binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P083635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124524 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083635 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083635 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P083635 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P468153 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P203443 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58166 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84519 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19849 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53891 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28482 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53951 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97138 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27041 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83086 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97853 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9691 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80157 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73682 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30836 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56736 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56820 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33242 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71790 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65768 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92052 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53064 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32059 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19641 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97157 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10782465 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782456 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782460 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782455 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782464 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782461 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782462 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782458 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782466 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782459 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782457 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10782454 | - | - |
Rat Genome 230 2.0 Array | 1387983_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370474_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387983_at | - | - |
Rat Genome 230 2.0 Array | 1370474_at | - | - |
Affymetrix HT Rat Focus Array | 1370474_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370474_at | - | - |
Affymetrix HT Rat Focus Array | 1387983_at | - | - |
Affymetrix HT Rat Focus Array | 1387983_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500008356.rn.1 | TC1500001323.rn.1 | RTA TC |
Rat Expression Set 230 | 1387983_at | - | - |
Rat Expression Set 230 | 1370474_PM_at | - | - |
Rat Expression Set 230 | 1387983_PM_at | - | - |
Rat Expression Set 230 | 1370474_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782454 | - | - |
Rat Genome U34 Set | J03819_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21570656 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359879 | - | - |
RatRef-12 Expression BeadChip | ILMN_58243 | - | - |
