Assay Details
Gene Symbol: Arnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012781.3 | NP_036913.3 | 17 - 18 | 1990 | 63 | |
XM_006229494.2 | XP_006229556.1 | 18 - 19 | 2706 | 63 | ||
XM_006229495.2 | XP_006229557.1 | 17 - 18 | 2799 | 63 | ||
XM_006229496.2 | XP_006229558.1 | 17 - 18 | 3260 | 63 | ||
XM_006229497.2 | XP_006229559.1 | 17 - 18 | 2643 | 63 | ||
XM_006229498.2 | XP_006229560.1 | 17 - 18 | 1941 | 63 | ||
XM_008759602.1 | XP_008757824.1 | 17 - 18 | 2359 | 63 | ||
XM_008759603.1 | XP_008757825.1 | 17 - 18 | 2348 | 63 | ||
XM_008759604.1 | XP_008757826.1 | 17 - 18 | 5079 | 63 | ||
XM_008759605.1 | XP_008757827.1 | 17 - 18 | 2273 | 63 | ||
XM_008759606.1 | XP_008757828.1 | 17 - 18 | 2498 | 63 | ||
XM_008759607.1 | XP_008757829.1 | 17 - 18 | 2055 | 63 | ||
XM_008759608.1 | XP_008757830.1 | 16 - 17 | 2155 | 63 | ||
GenBank mRNA | AY264366.1 | - | 17 - 18 | 1919 | 63 | |
DQ223732.1 | - | 1990 | 63 | |||
U61157.1 | - | 17 - 18 | 2014 | 63 | ||
U61405.1 | - | 17 - 18 | 1990 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
in utero embryonic development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
central nervous system development
brain development
positive regulation of cell proliferation
response to estradiol
negative regulation of apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
aryl hydrocarbon receptor binding
protein heterodimerization activity
protein dimerization activity
response to hypoxia
in utero embryonic development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
central nervous system development
brain development
positive regulation of cell proliferation
response to estradiol
negative regulation of apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
aryl hydrocarbon receptor binding
protein heterodimerization activity
protein dimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P368731 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P591370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96063 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P591370 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P368731 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P368731 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P591370 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P368731 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P591370 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P540669 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58589 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95994 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35696 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 87154 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35723 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21004 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82678 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58135 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3870 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34752 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42705 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1216 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68103 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37373 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10723431 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723427 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723424 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723436 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723429 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723418 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723417 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723439 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723426 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723428 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723433 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723430 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723438 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723440 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723423 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723420 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723437 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723422 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723425 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723435 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723421 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723434 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723432 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10723419 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10723413 | - | - |
Rat Genome 230 2.0 Array | 1370611_at | - | - |
Rat Genome 230 2.0 Array | 1370611_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373315_at | - | - |
Rat Genome 230 2.0 Array | 1373315_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370611_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370611_at | - | - |
Affymetrix HT Rat Focus Array | 1373315_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373315_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100047457.rn.1 | TC0100005952.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100047455.rn.1 | TC0100005952.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100047453.rn.1 | TC0100005952.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100047454.rn.1 | TC0100005952.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100047452.rn.1 | TC0100005952.rn.1 | RTA TC |
Rat Expression Set 230 | 1370611_PM_at | - | - |
Rat Expression Set 230 | 1373315_PM_at | - | - |
Rat Expression Set 230 | 1373315_at | - | - |
Rat Expression Set 230 | 1370611_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723419 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723413 | - | - |
Rat Genome U34 Set | U61405_at | - | - |
Rat Genome U34 Set | rc_AI176425_at | - | - |
Rat Genome U34 Set | U61405_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22253954 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351096 | - | - |
RatRef-12 Expression BeadChip | ILMN_58639 | - | - |
