Assay Details
| Gene Symbol: Kcna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012970.3 | NP_037102.1 | 1 - 2 | 97 | 76 | |
| XM_006233135.2 | XP_006233197.1 | 1 - 2 | 101 | 76 | ||
| GenBank mRNA | J04731.1 | - | 1 - 2 | 97 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the J04731.1, NM_012970.3, XM_006233135.2 transcripts.
Set Membership: |
|

Kcna2
Genomic Map