Assay Details
Gene Symbol: Sag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013023.2 | NP_037155.2 | 11 - 12 | 1069 | 86 | |
XM_008767196.1 | XP_008765418.1 | 11 - 12 | 1332 | 86 | ||
GenBank mRNA | M60737.1 | - | 10 - 11 | 960 | 86 | |
X15353.1 | - | 10 - 11 | 960 | 86 | ||
X51781.1 | - | 11 - 12 | 1069 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P054775 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P018317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141743 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054775 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P018317 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054775 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P018317 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P018317 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P054775 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P374851 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700301711 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48474 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10448 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29760 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700301711 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10925112 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925117 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925124 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925125 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925121 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925114 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925113 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925122 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925123 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925119 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925118 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925115 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925120 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925126 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925116 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10925111 | - | - |
Rat Genome 230 2.0 Array | 1368112_at | - | - |
Rat Genome 230 2.0 Array | 1368112_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368112_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368112_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0900009297.rn.1 | TC0900001086.rn.1 | RTA TC |
Rat Expression Set 230 | 1368112_at | - | - |
Rat Expression Set 230 | 1368112_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10925111 | - | - |
Rat Genome U34 Set | X51781cds_s_at | - | - |
Rat Genome U34 Set | M60737_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21251635 | - | - |
ABI Rat Genome Survey Microarray | 21386418 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361232 | - | - |
RatRef-12 Expression BeadChip | ILMN_58249 | - | - |
