Assay Details
| Gene Symbol: Ccl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013025.2 | NP_037157.2 | 1 - 2 | 142 | 76 | |
| GenBank mRNA | EF121994.1 | - | 1 - 2 | 73 | 76 | |
| EF121995.1 | - | 1 - 2 | 73 | 76 | ||
| U06435.1 | - | 1 - 2 | 73 | 76 | ||
| U22414.1 | - | 1 - 2 | 142 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl3
Genomic Map