Assay Details
| Gene Symbol: Slc1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013032.3 | NP_037164.3 | 1 - 2 | 178 | 92 | |
| GenBank mRNA | BC061743.1 | - | 1 - 2 | 178 | 5599027 | 92 |
| D63772.1 | - | 209 | 92 | |||
| U21104.1 | - | 1 - 2 | 190 | 92 | ||
| U39555.1 | - | 1 - 2 | 268 | 92 | ||
| X94255.1 | - | 1 - 2 | 235 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a1
Genomic Map