Assay Details
| Gene Symbol: Tnr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013045.1 | NP_037177.1 | 19 - 20 | 3945 | 75 | |
| GenBank mRNA | Z18630.1 | - | 19 - 20 | 3945 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
neuron cell-cell adhesion
negative regulation of cell adhesion
nervous system development
associative learning
negative regulation of neuron projection development
telencephalon cell migration
negative regulation of cell-cell adhesion
extracellular matrix organization
negative regulation of axon extension
locomotory exploration behavior
negative regulation of axon extension involved in regeneration
modulation of synaptic transmission
negative regulation of synaptic transmission
synapse organization
neuromuscular process controlling balance
positive regulation of synaptic transmission, glutamatergic
positive regulation of transmission of nerve impulse
long-term synaptic potentiation
integrin binding
cell adhesion
neuron cell-cell adhesion
negative regulation of cell adhesion
nervous system development
associative learning
negative regulation of neuron projection development
telencephalon cell migration
negative regulation of cell-cell adhesion
extracellular matrix organization
negative regulation of axon extension
locomotory exploration behavior
negative regulation of axon extension involved in regeneration
modulation of synaptic transmission
negative regulation of synaptic transmission
synapse organization
neuromuscular process controlling balance
positive regulation of synaptic transmission, glutamatergic
positive regulation of transmission of nerve impulse
long-term synaptic potentiation
integrin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P147644 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P147649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161236 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P147649 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P147644 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P147644 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P147649 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P147649 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P147644 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11678 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16071 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13299 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69508 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66261 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33975 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59216 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45821 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10764959 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764957 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764968 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764965 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764967 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764961 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764964 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764948 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764954 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764971 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764958 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764956 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764949 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764963 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764969 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764950 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764970 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764951 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764960 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764962 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764966 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764953 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764952 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764955 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10764947 | - | - |
| Rat Genome 230 2.0 Array | 1369692_at | - | - |
| Rat Genome 230 2.0 Array | 1369692_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369692_at | - | - |
| Affymetrix HT Rat Focus Array | 1369692_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300004064.rn.1 | TC1300000587.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369692_PM_at | - | - |
| Rat Expression Set 230 | 1369692_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764947 | - | - |
| Rat Genome U34 Set | Z18630_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21077999 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353848 | - | - |
| RatRef-12 Expression BeadChip | ILMN_64583 | - | - |

Tnr
Genomic Map