Assay Details
| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013101.3 | NP_037233.3 | 1 - 2 | 395 | 77 | |
| GenBank mRNA | L27057.1 | - | 1 - 2 | 395 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde4a
Genomic Map