Assay Details
Gene Symbol: Cpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013128.1 | NP_037260.1 | 6 - 7 | 1197 | 92 | |
GenBank mRNA | BC085762.1 | - | 6 - 7 | 1196 | 7193151 | 92 |
J04625.1 | - | 6 - 7 | 1152 | 92 | ||
X51406.1 | - | 6 - 7 | 1197 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
