Assay Details
Gene Symbol: Cald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013146.2 | NP_037278.1 | 4 - 5 | 902 | 98 | |
XM_006236260.2 | XP_006236322.1 | 4 - 5 | 2452 | 98 | ||
XM_006236261.2 | XP_006236323.1 | 5 - 6 | 973 | 98 | ||
XM_006236262.2 | XP_006236324.1 | 5 - 6 | 1126 | 98 | ||
XM_008762763.1 | XP_008760985.1 | 6 - 7 | 1054 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P085851 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P276276 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P085846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143048 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085846 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085851 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P276276 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085851 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P276276 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085846 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P276276 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P085851 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P085846 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P660244 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P660246 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P315562 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18194 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92174 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74410 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80944 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1256 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10854448 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854450 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854459 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854454 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854458 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854461 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854455 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854453 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854456 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854457 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854451 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854462 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854460 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854452 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854449 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10854446 | - | - |
Rat Genome 230 2.0 Array | 1368824_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368824_at | - | - |
Rat Genome 230 2.0 Array | 1371969_at | - | - |
Rat Genome 230 2.0 Array | 1368823_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368823_at | - | - |
Rat Genome 230 2.0 Array | 1371969_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368823_at | - | - |
Affymetrix HT Rat Focus Array | 1368823_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368824_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368824_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400004436.rn.1 | TC0400000625.rn.1 | RTA TC |
Rat Expression Set 230 | 1368824_PM_at | - | - |
Rat Expression Set 230 | 1368824_at | - | - |
Rat Expression Set 230 | 1368823_at | - | - |
Rat Expression Set 230 | 1368823_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854446 | - | - |
Rat Genome U34 Set | rc_AI180288_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21155811 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357776 | - | - |
RatRef-12 Expression BeadChip | ILMN_48416 | - | - |
