Assay Details
Gene Symbol: Ass1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013157.3 | NP_037289.1 | 6 - 7 | 582 | 80 | |
XM_006233911.2 | XP_006233973.1 | 8 - 9 | 733 | 80 | ||
GenBank mRNA | BC063146.1 | - | 8 - 9 | 723 | 6888536 | 80 |
M36708.1 | - | 6 - 7 | 582 | 80 | ||
X12459.1 | - | 6 - 7 | 582 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
urea cycle
citrulline metabolic process
argininosuccinate metabolic process
kidney development
liver development
arginine biosynthetic process
aspartate metabolic process
acute-phase response
midgut development
aging
response to nutrient
response to toxic substance
response to zinc ion
response to mycotoxin
response to amine
response to estradiol
response to lipopolysaccharide
response to drug
response to amino acid
response to peptide hormone
positive regulation of nitric oxide biosynthetic process
response to steroid hormone
response to glucocorticoid
response to growth hormone
diaphragm development
response to fatty acid
cellular response to lipopolysaccharide
cellular response to amino acid stimulus
cellular response to ammonium ion
cellular response to cAMP
cellular response to interferon-gamma
cellular response to tumor necrosis factor
cellular response to glucagon stimulus
cellular response to oleic acid
cellular response to amine stimulus
cellular response to laminar fluid shear stress
cellular response to dexamethasone stimulus
negative regulation of leukocyte cell-cell adhesion
argininosuccinate synthase activity
ATP binding
toxic substance binding
amino acid binding
poly(A) RNA binding
urea cycle
citrulline metabolic process
argininosuccinate metabolic process
kidney development
liver development
arginine biosynthetic process
aspartate metabolic process
acute-phase response
midgut development
aging
response to nutrient
response to toxic substance
response to zinc ion
response to mycotoxin
response to amine
response to estradiol
response to lipopolysaccharide
response to drug
response to amino acid
response to peptide hormone
positive regulation of nitric oxide biosynthetic process
response to steroid hormone
response to glucocorticoid
response to growth hormone
diaphragm development
response to fatty acid
cellular response to lipopolysaccharide
cellular response to amino acid stimulus
cellular response to ammonium ion
cellular response to cAMP
cellular response to interferon-gamma
cellular response to tumor necrosis factor
cellular response to glucagon stimulus
cellular response to oleic acid
cellular response to amine stimulus
cellular response to laminar fluid shear stress
cellular response to dexamethasone stimulus
negative regulation of leukocyte cell-cell adhesion
argininosuccinate synthase activity
ATP binding
toxic substance binding
amino acid binding
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P447373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133620 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P447373 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P447373 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P447373 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P447373 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88560 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25173 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57020 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10835359 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835362 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835357 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835356 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835363 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835369 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835368 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835365 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835360 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835364 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835366 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835370 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835358 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835361 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835367 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10835355 | - | - |
Rat Genome 230 2.0 Array | 1370964_at | - | - |
Rat Genome 230 2.0 Array | 1370964_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370964_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370964_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300021859.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021865.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021867.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021872.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021858.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021862.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021868.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021870.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021869.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021878.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021879.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021874.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021863.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021873.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021861.rn.1 | TC0300002853.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300021866.rn.1 | TC0300002853.rn.1 | RTA TC |
Rat Expression Set 230 | 1370964_PM_at | - | - |
Rat Expression Set 230 | 1370964_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835355 | - | - |
Rat Genome U34 Set | X12459_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21671442 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361110 | - | - |
RatRef-12 Expression BeadChip | ILMN_51487 | - | - |
