Assay Details
Gene Symbol: Gucy2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013170.1 | NP_037302.1 | 11 - 12 | 1371 | 81 | |
GenBank mRNA | M55636.1 | - | 11 - 12 | 1371 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P837219 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P046614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81467 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P046614 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P837219 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P837219 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P046614 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P046614 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P837219 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P837219 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25876 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21906 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19107 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4388 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29259 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27115 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7345 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68137 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10866478 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866481 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866484 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866485 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866483 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866473 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866475 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866474 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866476 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866477 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866479 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866482 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866480 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866472 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10866471 | - | - |
Rat Genome 230 2.0 Array | 1369162_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369162_at | - | - |
Affymetrix HT Rat Focus Array | 1369162_at | - | - |
Affymetrix HT Rat Focus Array | 1369162_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400028145.rn.1 | TC0400003967.rn.1 | RTA TC |
Rat Expression Set 230 | 1369162_at | - | - |
Rat Expression Set 230 | 1369162_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866471 | - | - |
Rat Genome U34 Set | M55636_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21271326 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376343 | - | - |
