Assay Details
| Gene Symbol: Prkar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013181.1 | NP_037313.1 | 2 - 3 | 273 | 84 | |
| XM_008768311.1 | XP_008766533.1 | 2 - 3 | 329 | 84 | ||
| GenBank mRNA | M17086.1 | - | 2 - 3 | 273 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkar1a
Genomic Map