Assay Details
Gene Symbol: Mc5r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013182.2 | NP_037314.1 | 1 | 40 | 78 | |
GenBank mRNA | L27081.1 | - | 1 | 252 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the L27081.1, NM_013182.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
