Assay Details
Gene Symbol: Il6r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017020.3 | NP_058716.2 | 7 - 8 | 1003 | 94 | |
XM_006232587.2 | XP_006232649.1 | 7 - 8 | 2437 | 94 | ||
XM_006232588.2 | XP_006232650.1 | 7 - 8 | 2436 | 94 | ||
GenBank mRNA | M58587.1 | - | 7 - 8 | 1092 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell proliferation
response to gamma radiation
positive regulation of activation of Janus kinase activity
cell growth
cytokine-mediated signaling pathway
endocrine pancreas development
response to lipopolysaccharide
negative regulation of interleukin-8 production
positive regulation of chemokine production
positive regulation of interleukin-6 production
negative regulation of collagen biosynthetic process
response to cytokine
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of MAPK cascade
response to peptide hormone
response to ethanol
positive regulation of JAK-STAT cascade
positive regulation of smooth muscle cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
positive regulation of NF-kappaB transcription factor activity
response to glucocorticoid
response to cAMP
interleukin-6-mediated signaling pathway
ciliary neurotrophic factor-mediated signaling pathway
ciliary neurotrophic factor receptor activity
interleukin-6 receptor activity
interleukin-6 receptor binding
enzyme binding
interleukin-6 binding
protein homodimerization activity
ciliary neurotrophic factor binding
positive regulation of cell proliferation
response to gamma radiation
positive regulation of activation of Janus kinase activity
cell growth
cytokine-mediated signaling pathway
endocrine pancreas development
response to lipopolysaccharide
negative regulation of interleukin-8 production
positive regulation of chemokine production
positive regulation of interleukin-6 production
negative regulation of collagen biosynthetic process
response to cytokine
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of MAPK cascade
response to peptide hormone
response to ethanol
positive regulation of JAK-STAT cascade
positive regulation of smooth muscle cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
positive regulation of NF-kappaB transcription factor activity
response to glucocorticoid
response to cAMP
interleukin-6-mediated signaling pathway
ciliary neurotrophic factor-mediated signaling pathway
ciliary neurotrophic factor receptor activity
interleukin-6 receptor activity
interleukin-6 receptor binding
enzyme binding
interleukin-6 binding
protein homodimerization activity
ciliary neurotrophic factor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P772157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113907 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P772157 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P772157 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P772157 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P772157 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701880917 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27945 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100568 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102350 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76940 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701880917 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10824531 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824534 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824535 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824538 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824532 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824537 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824533 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824536 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824540 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824539 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10824530 | - | - |
Rat Genome 230 2.0 Array | 1386987_PM_at | - | - |
Rat Genome 230 2.0 Array | 1386987_at | - | - |
Affymetrix HT Rat Focus Array | 1386987_at | - | - |
Affymetrix HT Rat Focus Array | 1386987_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200027797.rn.1 | TC0200004086.rn.1 | RTA TC |
Rat Expression Set 230 | 1386987_at | - | - |
Rat Expression Set 230 | 1386987_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824530 | - | - |
Rat Genome U34 Set | M58587_at | - | - |
Rat Genome U34 Set | M58587_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20851835 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349175 | - | - |
RatRef-12 Expression BeadChip | ILMN_64145 | - | - |
