Assay Details
| Gene Symbol: Sod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017050.1 | NP_058746.1 | 1 - 2 | 165 | 62 | |
| GenBank mRNA | BC082800.1 | - | 1 - 2 | 145 | 7304983 | 62 |
| FQ209495.1 | - | 1 - 2 | 166 | 62 | ||
| FQ209941.1 | - | 1 - 2 | 161 | 62 | ||
| FQ210282.1 | - | 1 - 2 | 166 | 62 | ||
| M21060.1 | - | 1 - 2 | 68 | 62 | ||
| X05634.1 | - | 1 - 2 | 64 | 62 | ||
| Y00404.1 | - | 1 - 2 | 165 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sod1
Genomic Map