Assay Details
| Gene Symbol: Gabra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017069.3 | NP_058765.3 | 1 - 2 | 189 | 66 | |
| XM_006229511.2 | XP_006229573.1 | 1 - 2 | 301 | 66 | ||
| GenBank mRNA | X51991.1 | - | 1 - 2 | 196 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_017069.3, X51991.1 transcripts.
Set Membership: |
|

Gabra3
Genomic Map