Assay Details
Gene Symbol: Cps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017072.1 | NP_058768.1 | 33 - 34 | 4004 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
urea cycle
liver development
glycogen catabolic process
proteolysis
arginine biosynthetic process
glutamine catabolic process
midgut development
response to toxic substance
response to zinc ion
response to amine
triglyceride catabolic process
response to food
response to lipopolysaccharide
response to glucagon
response to oleic acid
response to drug
response to starvation
response to amino acid
cellular response to fibroblast growth factor stimulus
positive regulation of vasodilation
nitric oxide metabolic process
response to steroid hormone
homocysteine metabolic process
response to glucocorticoid
response to cAMP
anion homeostasis
response to growth hormone
hepatocyte differentiation
carbamoyl phosphate biosynthetic process
cellular response to cAMP
cellular response to glucagon stimulus
cellular response to oleic acid
response to dexamethasone
carbamoyl-phosphate synthase (ammonia) activity
carbamoyl-phosphate synthase (glutamine-hydrolyzing) activity
endopeptidase activity
calcium ion binding
ATP binding
phospholipid binding
glutamate binding
protein complex binding
modified amino acid binding
urea cycle
liver development
glycogen catabolic process
proteolysis
arginine biosynthetic process
glutamine catabolic process
midgut development
response to toxic substance
response to zinc ion
response to amine
triglyceride catabolic process
response to food
response to lipopolysaccharide
response to glucagon
response to oleic acid
response to drug
response to starvation
response to amino acid
cellular response to fibroblast growth factor stimulus
positive regulation of vasodilation
nitric oxide metabolic process
response to steroid hormone
homocysteine metabolic process
response to glucocorticoid
response to cAMP
anion homeostasis
response to growth hormone
hepatocyte differentiation
carbamoyl phosphate biosynthetic process
cellular response to cAMP
cellular response to glucagon stimulus
cellular response to oleic acid
response to dexamethasone
carbamoyl-phosphate synthase (ammonia) activity
carbamoyl-phosphate synthase (glutamine-hydrolyzing) activity
endopeptidase activity
calcium ion binding
ATP binding
phospholipid binding
glutamate binding
protein complex binding
modified amino acid binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P346483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132012 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P346483 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P346483 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P346483 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P346483 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701734311 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103749 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95658 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33405 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101136 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14181 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70454 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 43390 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22862 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4683 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701734311 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10924094 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924116 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924091 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924124 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924121 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924115 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924109 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924093 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924108 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924112 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924104 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924105 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924111 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924096 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924098 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924125 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924101 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924114 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924095 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924097 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924126 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924118 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924103 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924110 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924090 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924100 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924122 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924092 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924113 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924123 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924120 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924107 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924117 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924106 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924089 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924099 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924119 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924102 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10924088 | - | - |
Rat Genome 230 2.0 Array | 1370151_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370151_at | - | - |
Affymetrix HT Rat Focus Array | 1370151_at | - | - |
Affymetrix HT Rat Focus Array | 1370151_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0900006884.rn.1 | TC0900000831.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900006883.rn.1 | TC0900000831.rn.1 | RTA TC |
Rat Expression Set 230 | 1370151_PM_at | - | - |
Rat Expression Set 230 | 1370151_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924088 | - | - |
Rat Genome U34 Set | M11710cds_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21829067 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1649842 | - | - |
